Learn More
anti-Palladin, Alexa Fluor 647, Clone: 1E6, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP125959AF647
Catalog No. NBP125959Y
Add to cart
Description
Description
Palladin Monoclonal antibody specifically detects Palladin in Human, Mouse, Rat, Canine, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
Palladin
1000000
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Q8WX93
CGI151, CGI-151, FLJ22190, FLJ38193, KIAA0992FLJ39139, MYN, myoneurin, palladin, palladin, cytoskeletal associated protein, PNCA1, Sarcoma antigen NY-SAR-77, SIH002FLJ61376
Mouse
IgG1
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Chicken, Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
Purified
PALLD
Endogenous Palladin immunoprecipitated from chicken embryo fibroblasts.
85 kDa
0.1 ml
Cellular Markers, Cytoskeleton Markers, Signal Transduction
Primary
23022