Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-pan Cadherin, Clone: PC-79, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP141361

Catalog No. NBP141361

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

pan Cadherin
PC-79
Unconjugated
Lyophilized from 1.2% sodium acetate, with 2mg BSA and 0.01mg NaN3 as preservative. with 0.01 mg Sodium Azide
Affinity Purified
CDH1
Synthetic peptide corresponding to the C-terminal amino acids of chicken N-Cadherin with an extra N-terminal lysine residue (24amino acids) coupled to KLH.
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Snake.
Human, Mouse, Rat, Chicken, Rabbit, Reptile
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
LYOPH
Western Blot 1-2 ug/ml, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 2-4 ug/ml
P08641
Arc-1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDHE, cell-CAM 120/80, ECAD, E-cadherin, Epithelial cadherin, LCAM, UVOE-Cadherin, uvomorulin
Mouse
IgG1
0.1 mg
Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Plasma Membrane Markers, Signal Transduction
Primary
999
Reconstitute with deionized water.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only