Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-pan Cytokeratin, PE, Clone: C-11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP148348PE

Catalog No. NBP148348PE


Add to cart

Description

Description

Cytokeratin, pan Monoclonal antibody specifically detects Cytokeratin, pan in Human, Mouse, Rat, Mammal samples. It is validated for Flow Cytometry, Flow (Intracellular).
Specifications

Specifications

pan Cytokeratin
C-11
Flow Cytometry, Flow (Intracellular)
Purified
Mouse
IgG1
0.1 ml
Cytoskeleton Markers, Protein Kinase, Signal Transduction
Primary
3848
Human, Mammalia, Mouse, Rat
Flow Cytometry
PE
PBS with 0.05% Sodium Azide
KRT1
Keratin-enriched preparation from human epidermoid carcinoma cell line A431.
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
This Mouse monoclonal Cytokeratin (Pan-reactive) antibody reacts with Cytokeratin peptides 4, 5, 6, 8, 10, 13, 18. Cytokeratins are a member of intermediate filaments subfamily represented in epithelial tissues.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only