Learn More
Park7/DJ-1 Mouse anti-Human, DyLight 350, Clone: 4H4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP192715UV
Catalog No. NBP192715UV
Add to cart
Description
Description
Park7/DJ-1 Monoclonal antibody specifically detects Park7/DJ-1 in Human samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown.
Specifications
Specifications
Park7/DJ-1
4H4
Western Blot, Immunocytochemistry/Immunofluorescence
Q99497
DJ-1, DJ1FLJ34360, EC 3.4, FLJ27376, FLJ92274, Oncogene DJ1, Parkinson disease (autosomal recessive, early onset) 7, Parkinson disease protein 7, protein DJ-1
Mouse
IgG1
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Bovine and Mouse.
Immunocytochemistry, Immunofluorescence, Western Blot
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
Purified
PARK7
Full length recombinant human Park7(DJ-1) expressed in and purified from E. coli. [UniProt# Q99497]
21 kDa
0.1 ml
Alzheimers Research, Apoptosis, Cancer, DNA replication Transcription Translation and Splicing, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Tumor Suppressors
Primary
11315
Human