Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Park7/DJ-1 Antibody (4H4), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP192715F
Description
Park7/DJ-1 Monoclonal specifically detects Park7/DJ-1 in Human, Bovine, Mouse (Negative), Rat (Negative) samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated.Specifications
Park7/DJ-1 | |
Monoclonal | |
1mg/mL | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated | |
DJ-1, DJ1FLJ34360, EC 3.4, FLJ27376, FLJ92274, Oncogene DJ1, Parkinson disease (autosomal recessive, early onset) 7, Parkinson disease protein 7, protein DJ-1 | |
Mouse | |
21 kDa | |
0.1 mL | |
Alzheimers Research, Apoptosis, Cancer, DNA replication Transcription Translation and Splicing, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Tumor Suppressors | |
11315 | |
Human, Bovine, Mouse (Negative), Rat (Negative) | |
Purified |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoassay | |
4H4 | |
FITC | |
Q99497 | |
PARK7 | |
Full length recombinant human Park7(DJ-1) expressed in and purified from E. coli. [UniProt# Q99497] | |
Protein G purified | |
RUO | |
Primary | |
Human, Bovine and Mouse. | |
Store at 4C in the dark. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only