Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PARL, Clone: 8C4B2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP151546

Catalog No. NBP151546


Add to cart

Description

Description

PARL Monoclonal antibody specifically detects PARL in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

PARL
8C4B2
Western Blot 1:500-1:2000, ELISA 1:10000
Ascites
PARL
Purified recombinant fragment of PARL (aa112-167) expressed in E. Coli.
Unpurified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
ELISA, Western Blot
Unconjugated
Ascites with 0.03% Sodium Azide
EC 3.4.21.105, mitochondrial, presenilin associated, rhomboid-like, PRO2207, rhomboid 7 homolog 1
Mouse
IgG2b
0.1 ml
Alzheimers Research, Apoptosis, Mitochondrial Fusion Proteins, Neuroscience
Primary
55486
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only