Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PBEF/Visfatin/NAMPT, Clone: 1D3A12, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP128888

Catalog No. NBP128888

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

PBEF/Visfatin/NAMPT
1D3A12
Western Blot 1:500-1:2000, ELISA 1:10000
Ascites
NAMPT
Purified recombinant fragment of PBEF1(aa338-479) expressed in E. Coli.
Unpurified
RUO
Primary
10135
Human
Western Blot, ELISA
Unconjugated
Ascites with 0.03% Sodium Azide
EC 2.4.2.12, MGC117256, NAmPRTase, Nampt, nicotinamide phosphoribosyltransferase, PBEF1110035O14Rik, PBEF1DKFZp666B131, Pre-B cell-enhancing factor, pre-B-cell colony enhancing factor 1, Pre-B-cell colony-enhancing factor 1, VF, VISFATIN
Mouse
IgG1
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Specific for PBEF1.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only