Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PCGF1, Clone: RNL-4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP142519

Catalog No. NBP142519


Add to cart

Description

Description

PCGF1 Monoclonal antibody specifically detects PCGF1 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

PCGF1
RNL-4
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Supernatant
PCGF1
The antibody reacts with the NSP-C antigen.
Tissue culture supernatant
RUO
Store at 4C. Do not freeze.
Monoclonal
Recognizing epitopes in the neuro-endocrine specific proteins (NSP) C. This antibody was raised against the amino-terminal amino acid sequence of NSP-C. cross reactivity is observed with human and rat.
Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Tissue culture supernatant with 0.1% Sodium Azide
MGC10882, Nervous system Polycomb-1, NSPc1, NSPC1FLJ43754, polycomb group ring finger 1,2010002K04Rik, polycomb group RING finger protein 1, RING finger protein 68, RNF68RNF3A-2
Mouse
IgG1
0.2 ml
Zinc Finger
Primary
84759
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only