Learn More
anti-PCNA, Clone: 1157CT1.1.3, Novus Biologicals™
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232297
Catalog No. NBP232297
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
PCNA
1157CT1.1.3
Unconjugated
PBS with 0.09% Sodium Azide
Purified
PCNA
This PCNA antibody is generated from mice immunized with a KLH conjugated synthetic peptide between 236-261 amino acids from the C-terminal region of human PCNA.
28.769 kDa
400 ul
Autophagy, Base Excision Repair, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Polymerases, DNA Repair, Phospho Specific, Stem Cell Markers
Primary
5111
Western Blot, Immunohistochemistry
0.5 mg/mL
Western Blot 1:2000, Immunohistochemistry 1:10-1:500
P12004
cyclin, DNA polymerase delta auxiliary protein, MGC8367, proliferating cell nuclear antigen
Mouse
IgG1
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse, Rat