Learn More
anti-PD-1, DyLight 350, Clone: J116, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP143107UV
Catalog No. NBP143107UV
Add to cart
Description
Description
PD-1 Monoclonal antibody specifically detects PD-1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
PD-1
J116
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Affinity Purified
PDCD1
The immunogen for this antibody was PD1.
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Human
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2
Mouse
IgG1 Kappa
0.1 ml
Apoptosis, Stem Cell Markers
Primary
5133