Learn More
anti-Penicillin, Clone: 9, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP142176
Catalog No. NBP142176
Add to cart
Description
Description
Penicillin Monoclonal antibody specifically detects Penicillin in Non-species specific samples. It is validated for ELISA.
Specifications
Specifications
Penicillin
Pen 9
Unconjugated
0.1 mg/ml PBS with 0.05% Sodium Azide
Ampicillin, Benzylpenicillin, dicloxacillin, flucloxacillin, methicillin, PCN, Penicilin, Penicillin G, Phenoxymethylpenicillin
The antibody reacts with the Penicillin antigen.
Protein A or G purified
RUO
Primary
Pen 9 reacts mainly with the thiazolidin ring of penicillin, but not with the lactam ring. The nature of the side chain in the penicilloyl group does not effect antibody binding as was shown by testing Pen 9 against benzylpenicillin, ampicillin, amoxillin and 6-aminopenicillanic acid. The presence of carrier protein is not essential for the presentation of the antigen associated with Pen 9. Source: Pen 9 was raised in a BALB/c mouse against the benzylpenicilloyl group of penicillium, using a penicilloyl-protein (transferrin) conjugate. Splenocytes were fused with X63-Ag8.653 cells.
ELISA
0.1 mg/mL
ELISA 1:100 - 1:2000
Purified
Mouse
IgG1 Kappa
0.1 mg
Store at 4C. Do not freeze.
Monoclonal
Non-species specific