Learn More
anti-Peptide YY, DyLight 350, Clone: RPY-B12, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP150769UV
Catalog No. NBP150769UV
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Peptide YY
RPY-B12
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Affinity Purified
PYY
Synthetic peptide (full length, 34 amino acids)
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Rat
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
Peptide tyrosine tyrosine, peptide YY, PYY1, PYY-I
Mouse
IgG1
0.1 ml
Signal Transduction
Primary
5697