Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Periostin/OSF-2, Clone: 1A1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP147922

Catalog No. NBP147922


Add to cart

Description

Description

Periostin/OSF-2 Monoclonal antibody specifically detects Periostin/OSF-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Periostin/OSF-2
OTI1A1
Unconjugated
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide
Affinity Purified
POSTN
Human recombinant protein fragment corresponding to amino acids 1-230 of human Periostin/OSF-2 (NP_006466) produced in E.coli.
93.2 kDa
0.1 ml
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
This antibody will possibly detect all isoforms encoding the amino acid regions 1-230 of human Periostin/OSF-2.
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
1 mg/mL
Western Blot 1:500, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100
Q15063
MGC119510, MGC119511, OSF-2osteoblast specific factor 2 (fasciclin I-like), OSF2periodontal ligament-specific periostin, Osteoblast-specific factor 2, PDLPOSTN, periostin, periostin isoform thy2, periostin isoform thy4, periostin isoform thy6, periostin isoform thy8, periostin, osteoblast specific factor, PNRP11-412K4.1
Mouse
IgM
Affinity purified
RUO
Primary
10631
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only