Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PGLYRP4/PGRP-I beta Antibody (186C426) - Azide Free, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP233246
Description
PGLYRP4/PGRP-I beta Monoclonal specifically detects PGLYRP4/PGRP-I beta in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
PGLYRP4/PGRP-I beta | |
Monoclonal | |
0.5 mg/mL | |
Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (Uehara et al (2005)), Flow (Cell Surface) reported in scientific literature (Uehara et al (2005)) | |
peptidoglycan recognition protein 4, peptidoglycan recognition protein I beta, PGRP-I-beta | |
Mouse | |
Protein G purified | |
RUO | |
57115 | |
Human | |
Purified |
Western Blot, Immunocytochemistry, Immunofluorescence, Flow Cytometry | |
186C426 | |
Unconjugated | |
Q96LB8 | |
PGLYRP4 | |
This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 95-110 of human PGRP-1 beta. | |
0.1 mg | |
Primary | |
PGRP-1b (186C426) | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction