Learn More
anti-PGP9.5 / UCHL-1, HRP, Clone: SPM575, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP234808H
Catalog No. NBP234808H
Add to cart
Description
Description
UCH-L1/PGP9.5 Monoclonal antibody specifically detects UCH-L1/PGP9.5 in Human, Mouse, Rat, Porcine, Bovine, Canine, Guinea Pig, Rabbit, Sheep, Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry-Frozen.
Specifications
Specifications
UCH-L1/PGP9.5
SPM575
Western Blot, ELISA, Immunohistochemistry-Frozen
Purified
UCHL1
Native UchL1 (PGP9.5) protein from brain
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
This MAb reacts with a protein of 20-30kDa, identified as PGP9.5, also known as ubiquitin carboxyl-terminal hydrolase-1 (UchL1). Initially, PGP9.5 expression in normal tissues was reported in neurons and neuroendocrine cells but later it was found in distal renal tubular epithelium, spermatogonia, Leydig cells, oocytes, melanocytes, prostatic secretory epithelium, ejaculatory duct cells, epididymis, mammary epithelial cells, Merkel cells, and dermal fibroblasts. Furthermore, immunostaining for PGP9.5 has been shown in a wide variety of mesenchymal neoplasms as well. A mutation in PGP9.5 gene is believed to cause a form of Parkinson's disease.
ELISA, Immunohistochemistry (Frozen), Western Blot
HRP
PBS with No Preservative
EC 3.4.19.12, EC 6.-, Neuron cytoplasmic protein 9.5, PARK5, PGP 9.5, PGP9.5, PGP9.5Uch-L1, PGP95, ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase), ubiquitin carboxyl-terminal hydrolase isozyme L1, ubiquitin C-terminal hydrolase, Ubiquitin thioesterase L1, UCHL1, UCH-L1
Mouse
IgG2a Kappa
0.1 ml
Alzheimers Research, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary
7345
Bovine, Canine, Guinea Pig, Human, Mouse, Porcine, Rabbit, Rat, Sheep, Zebrafish