Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Phospho-Tyrosine, Clone: G104, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications

Manufacturer:  Novus Biologicals NBP147562

Catalog No. NBP147562


Add to cart

Description

Description

Phospho-Tyrosine Monoclonal specifically detects Phospho-Tyrosine in Non-species specific samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

Phospho-Tyrosine
G104
Unconjugated
PBS (pH 7.4) and 50% Glycerol with 0.01% Sodium Azide
2 amino 3(4 hydroxyphenyl) propanoic acid, 4 hydroxyphenylalanine, phosphotyrosine, pTyrosine, Tyrosine
Phosphotyrosine, alanine and glyceine in a 1:1:1 ratio polymerized in the presence of keyhole limpet hemocyanin with 1-ethyl-3-(3'-dimentrylaminopropyl) carbodiimide
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Nonspecies specific
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
1 mg/mL
Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Purified
Mouse
IgG1
0.1 ml
Neuroscience, Signal Transduction, Tyrosine Kinases
Primary
Reacts with phosphotyrosine, and detects the presence of phosphotyrosine in both un-stimulated and stimulated cell lysates. Does not cross-react with phosphoserine or phosphothreonine.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only