Learn More
anti-Plasma Cell, DyLight 650, Clone: LIV3G11 (7B18), Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP233162C
Catalog No. NBP233162C
Add to cart
Description
Description
Plasma Cell Monoclonal antibody specifically detects Plasma Cell in Human, Rat (Negative) samples. It is validated for Immunohistochemistry-Paraffin.
Specifications
Specifications
Plasma Cell
LIV3G11 (7B18)
Immunohistochemistry-Paraffin
Purified
Mouse
IgG2a
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Rat (Negative)
Immunohistochemistry (Paraffin)
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
Plasma
Pancreatic cancer related serum mucin was used as the immunogen for the Plasma Cell antibody.
Protein A purified
RUO
Primary
It recognizes an intra-cytoplasmic antigen, which shows a very high degree of specificity for plasma cells. This antigen is present in normal as well as neoplastic plasma cells. Plasma cells, which are large lymphocytes derived from an antigen-specific B cell, secrete antibodies and are responsible for humoral immunity. Plasma cells differentiate from B cells upon stimulation by CD4+ lymphocytes. The B cell acts as an antigen-presenting cell (APC), consuming an offending pathogen, which is taken up by the B cell by phagocytosis and broken down within proteosomes. Plasma cells contain basophilic cytoplasm; their nucleus contains heterochromatin organized in a characteristic cartwheel arrangement. This MAb superbly recognizes normal and neoplastic plasma cells in routine formalin-fixed, paraffin-embedded tissue sections. It is of potential value in identifying myeloma or plasmacytoma in bone marrow or other tissues. It also helps differentiate lympho-plasmacytoid lymphoma from lymphocytic and follicular lymphoma. Note that this MAb is not suitable for staining frozen tissues.