Learn More
anti-Plasminogen, Clone: 4D2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB1107927
Catalog No. NB1107927
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Plasminogen Monoclonal antibody specifically detects Plasminogen in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
Plasminogen
4D2
ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Purified
PLG
Full length native protein (purified) (Human).
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Plasminogen (4D2)
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.4) with 0.09% Sodium Azide
DKFZp779M0222, EC 3.4.21, EC 3.4.21.7, plasmin, plasminogen
Mouse
IgG2b
0.2 mg
Cell Cycle and Replication
Primary
5340
Human