Learn More
anti-Presenilin-1, Clone: 34-4C3, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP178992
Catalog No. NBP178992
Add to cart
Description
Description
Presenilin-1 Monoclonal antibody specifically detects Presenilin-1 in Human, Mouse, Rat, Primate samples. It is validated for Immunohistochemistry, Immunohistochemistry-Frozen, Western Blot (Negative).
Specifications
Specifications
Presenilin-1
34-4C3
Unconjugated
PBS with 0.02% Sodium Azide
Purified
PSEN1
The I3 loop of the M2 receptor fusion protein (residues 225-359) was used as an immunogen
52.7 kDa
0.1 mg
Alzheimers Research, Apoptosis, Cancer, Hematopoietic Stem Cell Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission, Stem Cell Markers, Stem Cell Signaling Pathway, Tumor Suppressors, Wnt Signaling Pathway
Primary
5663
Human, Mouse, Primate, Rat
Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
1 mg/mL
Immunohistochemistry 1-3ug/ml, Immunohistochemistry-Frozen 1-3ug/ml, Western Blot (Negative)
P49768
AD3EC 3.4.23.-, Alzheimer disease 3, EC 3.4.23, FAD, presenilin 1, Protein S182, PS-1, PS1presenilin-1, PSNL1, S182
Mouse
IgG1
Protein A purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
It works well in human, rat, mouse and monkey.