Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BNP Mouse anti-Human, Clone: 3A6F7C7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP147509

Catalog No. NBP147509


Add to cart

Description

Description

BNP Monoclonal antibody specifically detects BNP in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

BNP
3A6F7C7
ELISA 1:10000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:1000
Ascites
NPPB
Synthetic peptide corresponding to aa (Glu-Pro-Leu-Gln-Glu-Ser-Pro-Arg-Pro-Thr-Gly-Val-Trp-Cys) of human BNP, conjugated to KLH.
Unpurified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Ascites with 0.03% Sodium Azide
BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide B, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein
Mouse
IgG1
0.1 ml
Cytokine Research, Immunology, Neuroscience, Neurotransmission
Primary
4879
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only