Learn More
anti-Progesterone R/NR3C3, DyLight 350, Clone: PR484, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP233325UV
Catalog No. NBP233325UV
Add to cart
Description
Description
Progesterone R/NR3C3 Monoclonal antibody specifically detects Progesterone R/NR3C3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
Progesterone R/NR3C3
PR484
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Purified
PGR
A progesterone receptor recombinant protein was used as the immunogen for the antibody. The specific epitope has not been mapped.
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
This MAb is specific to progesterone receptor and shows minimal cross-reaction with other members of the family. Progesterone receptor is expressed as two major isoforms, PR-A (81kDa) and PR-B (116kDa). Expression of PgR has been suggested to reflect a intact estrogen regulatory machinery and therefore, predict better clinical response to endocrine therapy than ER alone. It is excellent for immunohistochemical staining of formalin/paraffin tissues.
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
NR3C3PRNuclear receptor subfamily 3 group C member 3, progesterone receptor
Mouse
IgG1 Kappa
0.1 ml
Breast Cancer, Cancer
Primary
5241
Human