Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Progesterone R/NR3C3, PE, Clone: PR484, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP233325PE

Catalog No. NBP233325PE


Add to cart

Description

Description

Progesterone R/NR3C3 Monoclonal antibody specifically detects Progesterone R/NR3C3 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Progesterone R/NR3C3
PR484
Flow Cytometry
Purified
PGR
A progesterone receptor recombinant protein was used as the immunogen for the antibody. The specific epitope has not been mapped.
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
This MAb is specific to progesterone receptor and shows minimal cross-reaction with other members of the family. Progesterone receptor is expressed as two major isoforms, PR-A (81kDa) and PR-B (116kDa). Expression of PgR has been suggested to reflect a intact estrogen regulatory machinery and therefore, predict better clinical response to endocrine therapy than ER alone. It is excellent for immunohistochemical staining of formalin/paraffin tissues.
Flow Cytometry
PE
PBS with 0.05% Sodium Azide
NR3C3PRNuclear receptor subfamily 3 group C member 3, progesterone receptor
Mouse
IgG1 Kappa
0.1 ml
Breast Cancer, Cancer
Primary
5241
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only