Learn More
anti-Prohibitin, Clone: 1215CT487.109.106, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232305
Catalog No. NBP232305
Add to cart
Description
Description
Prohibitin Monoclonal antibody specifically detects Prohibitin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
Prohibitin
1215CT487.109.106
Unconjugated
PBS with 0.09% Sodium Azide
Purified
PHB
This antibody is generated from a mice immunized with a recombinant protein between 1-205 amino acids from human.
29.804 kDa
400 ul
Cell Cycle and Replication, Core ESC Like Genes, Ovarian Carcinoma Cell Markers, Stem Cell Markers
Primary
5245
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
0.5 mg/mL
Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:25
P35232, P35232
PHB1, prohibitin
Mouse
IgG2b Kappa
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse, Rat