Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRR7 Antibody (TRAP3/10), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP130424UV
Description
PRR7 Monoclonal specifically detects PRR7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
PRR7 | |
Monoclonal | |
DyLight 350 | |
MGC10772, proline rich 7 (synaptic), proline-rich protein 7, Synaptic proline-rich membrane protein | |
Mouse | |
Protein A purified | |
RUO | |
80758 | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunocytochemistry, Immunofluorescence | |
TRAP3/10 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
PRR7 | |
Recombinant C-terminal half of the intracellular domain of human PRR7/TRAP3 (amino acids 126-253) | |
0.1 mL | |
Primary | |
The mouse monoclonal antibody TRAP3/10 recognizes an epitope located in the C-terminal part of the intracellular domain of PRR7/TRAP3 (amino acids 126-253 of human PRR7 TRAP3), a 28 kDa proline-rich membrane protein presumably associated with NMDA receptor complex. | |
Store at 4C in the dark. | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction