Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PRR7, DyLight 405, Clone: TRAP3/10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP130424V

Catalog No. NBP130424V


Add to cart

Description

Description

PRR7 Monoclonal antibody specifically detects PRR7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

PRR7
TRAP3/10
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
PRR7
Recombinant C-terminal half of the intracellular domain of human PRR7/TRAP3 (amino acids 126-253)
Protein A purified
RUO
Primary
80758
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Western Blot
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
MGC10772, proline rich 7 (synaptic), proline-rich protein 7, Synaptic proline-rich membrane protein
Mouse
IgG2a
0.1 ml
Store at 4C in the dark.
Monoclonal
The mouse monoclonal antibody TRAP3/10 recognizes an epitope located in the C-terminal part of the intracellular domain of PRR7/TRAP3 (amino acids 126-253 of human PRR7 TRAP3), a 28 kDa proline-rich membrane protein presumably associated with NMDA receptor complex.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only