Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-R-Spondin 1, Clone: 7A6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP128867

Catalog No. NBP128867


Add to cart

Description

Description

R-Spondin 1 Monoclonal antibody specifically detects R-Spondin 1 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

R-Spondin 1
7A6
Western Blot 1:500-1:2000, ELISA 1:10000
Ascites
RSPO1
Purified recombinant fragment of RSPO1 expressed in E. Coli.
28.9 kDa
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Specific for R-spondin1.
ELISA, Western Blot
Unconjugated
Ascites with 0.03% Sodium Azide
CRISTIN3, FLJ40906Roof plate-specific spondin-1, hRspo1, roof plate-specific spondin, RSPO, RSPONDIN, R-spondin homolog (Xenopus laevis), R-spondin1, R-spondin-1
Mouse
IgG1
Unpurified
RUO
Primary
284654
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only