Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rad9 Antibody (OTI7B11), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP147946
Description
Rad9 Monoclonal specifically detects Rad9 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Rad9 | |
Monoclonal | |
2 mg/mL | |
Western Blot 1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50 | |
RAD9A | |
Full-length protein expressed in 293T cell transfected with human RAD9A expression vector | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is specific for Homo sapiens RAD9 homolog A (S. pombe) (RAD9A). | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
OTI7B11 | |
Unconjugated | |
cell cycle checkpoint control protein RAD9A, DNA repair exonuclease rad9 homolog A, EC 3.1.11.2, hRAD9, RAD9, RAD9 (S. pombe) homolog, RAD9 homolog A (S. pombe) | |
Mouse | |
42.5 kDa | |
0.1 mL | |
Apoptosis, Cancer, Checkpoint signaling, Chromatin Research, DNA Repair, Tumor Suppressors | |
5883 | |
Human | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction