Learn More
Caspase-12 Rabbit anti-Mouse, Rat, Polyclonal, R&D Systems™
Rabbit Polyclonal Antibody
Manufacturer:  R&D Systems AF1456
Catalog No. AF1456
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
Caspase-12 Polyclonal antibody AF1456 Specifically detects Caspase-12 in Mouse, Rat samples. It is validated for Western Blotting
Specifications
Specifications
Caspase-12
LYOPH
Western Blot 0.25 ug/mL
O08736
Apoptosis Related Cysteine Protease, CASP 12, casp12, CASP12P1, Caspase 12 pseudogene 1, Caspase12, UNQ9415
Rabbit
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects endogenous mouse and rat Casapase-12 proenzyme in Western blots.
Mouse, Rat
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
Casp12
E. coli-derived recombinant mouse Caspase-12 Met1-Asn419 (predicted) Accession # O08736
Immunogen affinity purified
RUO
Primary
100506742
Reconstitute at 0.2 mg/mL in sterile PBS.