Learn More
RBFOX3/NeuN Mouse anti-Human, Mouse, Rat, APC, Clone: 1B7, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP192693APC
Catalog No. NBP192693J
Add to cart
Description
Description
RBFOX3/NeuN Monoclonal antibody specifically detects RBFOX3/NeuN in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
Specifications
RBFOX3/NeuN
1B7
Flow Cytometry
A6NFN3
FLJ56884, FLJ58356, Fox-1 homolog C, FOX3, FOX-3, hexaribonucleotide binding protein 3, HRNBP3, NeuN, neuronal nuclei, RBFOX3, RNA binding protein fox-1 homolog 3, RNA binding protein, fox-1 homolog (C. elegans) 3
Mouse
IgG2a
0.1 ml
Neuronal Cell Markers, Neuroscience
Primary
146713
Human, Mouse, Rat
Flow Cytometry
APC
PBS with 0.05% Sodium Azide
Purified
RBFOX3
N-terminal 100 amino acids of human FOX3 as expressed in and purified from E. coli
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Bovine, Porcine, Mouse, Rat and other Mammals.