Learn More
RBFOX3/NeuN Mouse anti-Human, Mouse, Rat, DyLight 755, Clone: 1B7, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP192693IR
Catalog No. NBP192693IR
Add to cart
Description
Description
RBFOX3/NeuN Monoclonal antibody specifically detects RBFOX3/NeuN in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications
Specifications
RBFOX3/NeuN
1B7
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
A6NFN3
FLJ56884, FLJ58356, Fox-1 homolog C, FOX3, FOX-3, hexaribonucleotide binding protein 3, HRNBP3, NeuN, neuronal nuclei, RBFOX3, RNA binding protein fox-1 homolog 3, RNA binding protein, fox-1 homolog (C. elegans) 3
Mouse
IgG2a
0.1 ml
Neuronal Cell Markers, Neuroscience
Primary
146713
Human, Mouse, Rat
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
Purified
RBFOX3
N-terminal 100 amino acids of human FOX3 as expressed in and purified from E. coli
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Bovine, Porcine, Mouse, Rat and other Mammals.