Learn More
anti-Renal Cell Carcinoma (gp200), DyLight 488, Clone: HD30, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP141003G
Catalog No. NBP141003G
Add to cart
Description
Description
DEC-205/CD205 Monoclonal specifically detects DEC-205/CD205 in Human, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
Renal Cell Carcinoma (gp200)
HD30
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
O60449
CD205LY-75, CLEC13BLy-75, C-type lectin domain family 13 member B, DEC-205CD205 antigen, GP200-MR6, lymphocyte antigen 75
Mouse
IgG1
0.1 ml
Dendritic Cell Markers, Ovarian Carcinoma Cell Markers, Renal Cell Carcinoma Cell Markers, Vision
Primary
4065
Human, Primate
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
Purified
LY75
Purified extracellular domain of human CD205-IgG1 Fc fusion protein.
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
This antibody specifically recognises CD205, otherwise known as DEC205, a type I transmembrane glycoprotein and member of the macrophage mannose receptor family of C-type lectins, predominantly expressed by human dendritic cells.