Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Renal Cell Carcinoma (gp200), PE, Clone: HD30, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP141003PE

Catalog No. NBP141003PE


Add to cart

Description

Description

DEC-205/CD205 Monoclonal antibody specifically detects DEC-205/CD205 in Human, Primate samples. It is validated for Flow Cytometry.
Specifications

Specifications

Renal Cell Carcinoma (gp200)
HD30
Flow Cytometry
O60449
CD205LY-75, CLEC13BLy-75, C-type lectin domain family 13 member B, DEC-205CD205 antigen, GP200-MR6, lymphocyte antigen 75
Mouse
IgG1
0.1 ml
Dendritic Cell Markers, Ovarian Carcinoma Cell Markers, Renal Cell Carcinoma Cell Markers, Vision
Primary
4065
Human, Primate
Flow Cytometry
PE
PBS with 0.05% Sodium Azide
Purified
LY75
Purified extracellular domain of human CD205-IgG1 Fc fusion protein.
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
This antibody specifically recognises CD205, otherwise known as DEC205, a type I transmembrane glycoprotein and member of the macrophage mannose receptor family of C-type lectins, predominantly expressed by human dendritic cells.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only