Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-SCIMP, DyLight 488, Clone: NVL-07, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222349G
Description
SCIMP Monoclonal specifically detects SCIMP in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
SCIMP | |
Monoclonal | |
DyLight 488 | |
Q6UWF3 | |
SCIMP | |
Recombinant intracellular part of human SCIMP. | |
0.1 mL | |
Primary | |
The mouse monoclonal antibody NVL-07 recognizes intracellular part of human transmembrane adaptor SCIMP. This protein of 17 kDa predicted Mw migrates as a 22 kDa band on SDS PAGE. | |
Store at 4C in the dark. | |
IgG2 α |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
NVL-07 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
transmembrane protein C17orf87, C17orf87, chromosome 17 open reading frame 87, DTFT5783, FLJ32580, SLP adapter and CSK-interacting membrane protein, SLP adaptor and CSK interacting membrane protein, SLP65/SLP76, Csk-interacting membrane protein, UNQ5783 | |
Mouse | |
Protein A purified | |
RUO | |
388325 | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only