Learn More
anti-Semaphorin 4D/CD100, Clone: 133.1G6, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP142504
Catalog No. NBP142504
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Semaphorin 4D/CD100
133.1G6
Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500, CyTOF-ready
Purified
SEMA4D
The antibody reacts with the CD100 antigen.
IgM purified
RUO
Store at 4C. Do not freeze.
Monoclonal
133.1G6 reacts with human CD100, a novel 150 kD homodimer cell-surface antigen that is expressed on resting and PHA-stimulated T cells. The protein is weakly expressed on NK cells, EBV transformed B cells, monocytes and tumor T cell lines. The mAb was clustered at the IV WLDA (workshop no. A022). Source: A BALB/c mouse was immunized with PHA stimulated PBL. Splenocytes were fused with mouse myeloma NS1 cells.
Flow Cytometry, Immunoprecipitation, CyTOF
Unconjugated
0.1 mg/ml PBS with 0.05% Sodium Azide
A8, CD100 antigen, CD100M-sema-G, coll-4, FLJ33485, FLJ34282, FLJ39737, FLJ46484, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, 4D, MGC169138, MGC169141, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4D, semaphorin-4D
Mouse
IgM
0.1 mg
Apoptosis, Immunology
Primary
10507
Human, Mouse