Learn More
Serpin F2/alpha 2-Antiplasmin Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Manufacturer:  R&D Systems AF1239
Catalog No. AF1239
Add to cart
This item is not returnable. View return policy
Description
Description
Serpin F2/alpha 2-Antiplasmin Polyclonal specifically detects Serpin F2/alpha 2-Antiplasmin in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.
Specifications
Specifications
Serpin F2/alpha 2-Antiplasmin
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL
Q61247
AAPclade F (alpha-2 antiplasmin, alpha 2-Antiplasmin, Alpha-2-AP, Alpha-2-PI, Alpha-2-plasmin inhibitor, pigment epithelium derived factor), member 2, Serpin F2, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 2
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse Serpin F2/alpha 2 -Antiplasmin in direct ELISAs and Western blots. In Western blots, approximately 15% cross-reactivity with recombinant human (rh) Serpin F2 and less than 1% cross-reactivity with recombinant mouse (rm) Serpin C1, rmSerpin D1, rhSerpin A1, rmSerpin A5, and rhSerpin C1 is observed.
Mouse
Western Blot, Immunohistochemistry, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
SERPINF2
Mouse myeloma cell line NS0-derived recombinant mouse Serpin F2/alpha 2 -Antiplasmin Val28-Lys491 Accession # Q61247
Immunogen affinity purified
RUO
Primary
5345
Reconstitute at 0.2 mg/mL in sterile PBS.