Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SF3B4 Antibody (3A1), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP192692AF488
Description
SF3B4 Monoclonal specifically detects SF3B4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
SF3B4 | |
Monoclonal | |
Alexa Fluor 488 | |
Q15427 | |
SF3B4 | |
Full length recombinant human SF3B4 expressed in and purified from E. coli. [UniProt# Q15427] | |
Protein G purified | |
RUO | |
Primary | |
Human, Bovine, Porcine, Mouse, Rat and other Mammals. | |
Store at 4C in the dark. | |
IgG2b |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
3A1 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Hsh49, pre-mRNA splicing factor SF3b 49 kDa subunit, Pre-mRNA-splicing factor SF3b 49 kDa subunit, SAP 49, SAP49MGC10828, SF3B49, SF3b50, spliceosomal protein, spliceosome-associated protein (U2 snRNP), Spliceosome-associated protein 49, splicing factor 3B subunit 4, splicing factor 3b, subunit 4, 49kD, splicing factor 3b, subunit 4, 49kDa | |
Mouse | |
49 kDa | |
0.1 mL | |
Neuroscience | |
10262 | |
Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction