Learn More
SF3B4 Mouse anti-Human, Mouse, Rat, DyLight 755, Clone: 3A1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP192692IR
Catalog No. NBP192692IR
Add to cart
Description
Description
SF3B4 Monoclonal antibody specifically detects SF3B4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Specifications
Specifications
SF3B4
3A1
Western Blot, Immunocytochemistry/Immunofluorescence
Q15427
Hsh49, pre-mRNA splicing factor SF3b 49 kDa subunit, Pre-mRNA-splicing factor SF3b 49 kDa subunit, SAP 49, SAP49MGC10828, SF3B49, SF3b50, spliceosomal protein, spliceosome-associated protein (U2 snRNP), Spliceosome-associated protein 49, splicing factor 3B subunit 4, splicing factor 3b, subunit 4, 49kD, splicing factor 3b, subunit 4, 49kDa
Mouse
IgG2b
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Bovine, Porcine, Mouse, Rat and other Mammals.
Immunocytochemistry, Immunofluorescence, Western Blot
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
Purified
SF3B4
Full length recombinant human SF3B4 expressed in and purified from E. coli. [UniProt# Q15427]
49 kDa
0.1 ml
Neuroscience
Primary
10262
Human, Mouse, Rat