Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SF3B4 Mouse anti-Human, Mouse, Rat, FITC, Clone: 3A1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP192692F

Catalog No. NBP192692F


Add to cart

Description

Description

SF3B4 Monoclonal antibody specifically detects SF3B4 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

SF3B4
3A1
FITC
PBS with 0.05% Sodium Azide
Purified
SF3B4
Full length recombinant human SF3B4 expressed in and purified from E. coli. [UniProt# Q15427]
49 kDa
0.1 ml
Neuroscience
Primary
10262
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence
1mg/mL
Immunocytochemistry/Immunofluorescence
Q15427
Hsh49, pre-mRNA splicing factor SF3b 49 kDa subunit, Pre-mRNA-splicing factor SF3b 49 kDa subunit, SAP 49, SAP49MGC10828, SF3B49, SF3b50, spliceosomal protein, spliceosome-associated protein (U2 snRNP), Spliceosome-associated protein 49, splicing factor 3B subunit 4, splicing factor 3b, subunit 4, 49kD, splicing factor 3b, subunit 4, 49kDa
Mouse
IgG2b
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Bovine, Porcine, Mouse, Rat and other Mammals.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only