Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siglec-9 Mouse anti-Human, Canine, Monkey, Clone: OTI1D9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP147969

Catalog No. NBP147969


Add to cart

Description

Description

Siglec-9 Monoclonal antibody specifically detects Siglec-9 in Human, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

Siglec-9
OTI1D9
Unconjugated
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide
CD329, CDw329, FOAP-9, OBBP-LIKE, Protein FOAP-9, sialic acid binding Ig-like lectin 9, sialic acid-binding Ig-like lectin 9, siglec-9
Mouse
IgG1
Affinity purified
RUO
Primary
27180
Canine, Human, Monkey
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
0.7 mg/mL
Western Blot 1:20000-40000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50
Affinity Purified
SIGLEC9
Full length human recombinant protein of human SIGLEC9 (NP_055256) produced in HEK293T cell.
50.1 kDa
0.1 ml
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
This antibody is specific for Homo sapiens sialic acid binding Ig-like lectin 9 (SIGLEC9).
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only