Learn More
anti-Sirtuin 1/SIRT1, Clone: 1089CT5.3.1, Novus Biologicals™
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232309
Catalog No. NBP232309
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Sirtuin 1/SIRT1
1089CT5.3.1
Unconjugated
PBS with 0.09% Sodium Azide
Purified
SIRT1
This antibody is generated from a mice immunized with a recombinant protein between 1-180 amino acids from human.
81.681 kDa
400 ul
Apoptosis, Chromatin Research, DNA Repair, Epigenetics, Histone Deacetylases, Phospho Specific
Primary
23411
Western Blot
0.42 mg/mL
Western Blot 1:1000
Q96EB6
EC 3.5.1, S. cerevisiae, homolog) 1, sirtuin 1
Mouse
IgG3 Kappa
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human