Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-SLC6A3/DAT1, Alexa Fluor 405, Clone: mAb16, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222164AF405
Description
SLC6A3/DAT1 Monoclonal specifically detects SLC6A3/DAT1 in Mouse, Rat, Human (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SLC6A3/DAT1 | |
Monoclonal | |
Alexa Fluor 405 | |
DA transporter, DATDAT1Solute carrier family 6 member 3, sodium-dependent dopamine transporter, solute carrier family 6 (neurotransmitter transporter, dopamine), member 3 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Mouse and rat. Little to no human reactivity has been observed. | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
mAb16 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
SLC6A3 | |
Synthetic peptide corresponding to the N-terminus of rat DAT1. [UniProt# P23977] | |
0.1 mL | |
Neuroscience | |
6531 | |
Mouse, Rat, Human (Negative) | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only