Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Slug, Clone: 2H5, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP147486

Catalog No. NBP147486

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Slug
2H5
Western Blot 1:500-1:2000, ELISA 1:10000
Ascites
SNAI2
Purified recombinant fragment of human SNAI2 expressed in E. Coli.
30 kDa
0.1 ml
Neuronal Cell Markers, Neuronal Stem Cell Markers
Primary
6591
ELISA, Western Blot
Unconjugated
Ascites with 0.03% Sodium Azide
MGC10182, Neural crest transcription factor Slug, Protein snail homolog 2, slug homolog, zinc finger protein (chicken), SLUGH, SLUGzinc finger protein, snail 2, snail homolog 2 (Drosophila), WS2D, zinc finger protein SNAI2
Mouse
IgG1
Unpurified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only