Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Smallpox B5R, Clone: 1J2F3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP233255

Catalog No. NBP233255


Add to cart

Description

Description

Smallpox B5R Monoclonal antibody specifically detects Smallpox B5R in Virus samples. It is validated for Western Blot.
Specifications

Specifications

Smallpox B5R
1J2F3
Unconjugated
0.2 ml PBS - BSA free with No Preservative
Purified
Within the range of Amino acids 33-62 of the B5R protein were used as the immunogen.
Protein G purified
RUO
Primary
1486562
Virus
Western Blot
0.5 mg/mL
Western Blot 1-3 ug/ml
P33823
Mouse
IgG1 Kappa
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Monoclonal
The amino acid sequence used as immunogen for this phospho antibody is 100% homologous in Human, Chimpanzee, Dog, Rat, Mouse, Chicken and Zebrafish and 92% homologous in Xenopus.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only