Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-SOD1/Cu-Zn SOD, Clone: 72B1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP141974

Catalog No. NBP141974


Add to cart

Description

Description

SOD1/Cu-Zn SOD Monoclonal antibody specifically detects SOD1/Cu-Zn SOD in Human samples. It is validated for Immunoprecipitation.
Specifications

Specifications

SOD1/Cu-Zn SOD
72B1
Unconjugated
PBS and 50% Glycerol with No Preservative
ALS, ALS1, amyotrophic lateral sclerosis 1 (adult), Cu, Cu/Zn superoxide dismutase, EC 1.15.1.1, homodimer, hSod1, indophenoloxidase A, IPOA, SOD, SOD, soluble, superoxide dismutase [Cu-Zn], Superoxide dismutase 1, superoxide dismutase 1, soluble, Zn superoxide dismutase, EC 1.15.1.110superoxide dismutase, cystolic
Mouse
IgG1 Kappa
0.1 mg
Cancer, Diabetes Research, Mitochondrial Fusion Proteins, Neuronal Cell Markers, Neuroscience, Tumor Suppressors
Primary
6647
Immunoprecipitation
1 mg/mL
Immunoprecipitation 1:10-1:500
Affinity Purified
SOD1
Recombinant human protein purified from E.coli
Protein A or G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only