Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOD3/EC-SOD Mouse anti-Human, Mouse, Rat, Guinea Pig, Alexa Fluor 405, Clone: 4GG11G6, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP122417AF405
Description
SOD3/EC-SOD Monoclonal antibody specifically detects SOD3/EC-SOD in Human, Mouse, Rat, Guinea Pig samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SOD3/EC-SOD | |
4GG11G6 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
P08294 | |
EC-SODEC 1.15.1.1, extracellular superoxide dismutase [Cu-Zn], MGC20077, superoxide dismutase 3, extracellular | |
Mouse | |
IgG1 Kappa | |
0.1 ml | |
Store at 4C in the dark. | |
Monoclonal | |
Detects extracellular SOD approx 35kDa. |
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Purified | |
SOD3 | |
Human extracellular SOD purified from aortas | |
Protein G purified | |
RUO | |
Primary | |
6649 | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only