Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Sonic Hedgehog/Shh, Clone: 8G3, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP151615

Catalog No. NBP151615

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Sonic Hedgehog/Shh
8G3
Western Blot 1:200-1:1000, ELISA 1:10000
Ascites
SHH
Purified recombinant fragment of human SHH expressed in E. Coli.
49.6 kDa
0.1 ml
Neuroscience, Sensory Systems, Stem Cell Signaling Pathway, Stem Cells, Vision
Primary
6469
ELISA, Western Blot
Unconjugated
Ascites with 0.03% Sodium Azide
HHG1, HHG-1, HLP3, HPE3, MCOPCB5sonic hedgehog (Drosophila) homolog, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS
Mouse
IgG1
Unpurified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only