Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-SorLA, Clone: 7D7B11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP128885

Catalog No. NBP128885


Add to cart

Description

Description

SorLA Monoclonal antibody specifically detects SorLA in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

SorLA
7D7B11
Western Blot 1: 500-1: 2000, ELISA 1: 10000
Ascites
SORL1
Purified recombinant fragment of SORL1(aa2159-2214) expressed in E. Coli.
Unpurified
RUO
Primary
6653
Human
ELISA, Western Blot
Unconjugated
Ascites with 0.03% Sodium Azide
C11orf32, chromosome 11 open reading frame 32, FLJ21930, FLJ39258, gp250, Low-density lipoprotein receptor relative with 11 ligand-binding repeats, LR11LDLR relative with 11 ligand-binding repeats, LRP9, mosaic protein LR11, SorLA, SorLA-1SORLA, sortilin-related receptor, sortilin-related receptor, L(DLR class) A repeats containing, sortilin-related receptor, L(DLR class) A repeats-containing, Sorting protein-related receptor containing LDLR class A repeats
Mouse
IgG1
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Specific for SORL1.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only