Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Spectrin alpha 1, Clone: AF10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP142555

Catalog No. NBP142555


Add to cart

Description

Description

Spectrin alpha 1 Monoclonal antibody specifically detects Spectrin alpha 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications

Specifications

Spectrin alpha 1
AF10
Unconjugated
PBS solution containing 1.0% w/v BSA with 0.1% Sodium Azide
Purified
SPTA1
The antibody reacts with the Erythroid alpha-Spectrin antigen.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
The antibody is specific to the 240kD erythroid a-spectrin.
Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
0.1 mg/mL
Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
P02549
alpha-I spectrin, EL2, erythrocyte, HS3, spectrin, alpha, erythrocytic 1 (elliptocytosis 2)
Mouse
IgG1
0.05 mg
Cytoskeleton Markers
Primary
6708
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only