Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Src, Clone: 4F1E8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP147547

Catalog No. NBP147547


Add to cart

Description

Description

Src Monoclonal antibody specifically detects Src in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Src
4F1E8
Western Blot 1: 500 - 1:2000, ELISA 1:10000
Ascites
SRC
Purified recombinant fragment of SRC(aa10-193) expressed in E. Coli.
Unpurified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
ELISA, Western Blot
Unconjugated
Ascites with 0.03% Sodium Azide
c-src, EC 2.7.10, EC 2.7.10.2, pp60c-src, Rous sarcoma, tyrosine kinase pp60c-src, tyrosine-protein kinase SRC-1, v-src avian sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog, v-src sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog (avian)
Mouse
IgG2b
0.1 ml
Protein Kinase, Signal Transduction, Transcription Factors and Regulators, Tyrosine Kinases
Primary
6714
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only