Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAT1 Antibody (OTI4H9), Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP148011
Description
STAT1 Monoclonal specifically detects STAT1 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
STAT1 | |
Monoclonal | |
0.7 mg/mL | |
Western Blot 1:1000-2000, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:50-100, Immunohistochemistry-Paraffin 1:50 | |
STAT1 | |
Full length human recombinant protein of human STAT1 (NP_009330) produced in HEK293T cell. | |
Immunogen affinity purified | |
RUO | |
Primary | |
This antibody is specific for Homo sapiens signal transducer and activator of transcription 1, 91kDa (STAT1), transcript variant beta. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
OTI4H9 | |
Unconjugated | |
DKFZp686B04100, ISGF-3,91kD, signal transducer and activator of transcription 1, 91kDa, signal transducer and activator of transcription-1 | |
Mouse | |
82.9 kDa | |
0.1 mL | |
Apoptosis, Breast Cancer, Cancer, Signal Transduction, Transcription Factors and Regulators | |
6772 | |
Human, Mouse, Rat, Canine, Primate, Monkey | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction